Oxyde de zinc, nanopoudre, 99 % (pureté du métal), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxyde de zinc, nanopoudre, 99 % (pureté du métal), Thermo Scientific Chemicals
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
50 g
10 g
Référence 044533.18
également connu sous le numéro 044533-18
Prix (EUR)/ Each
414,00
Offre exceptionnelle en ligne
460.00 
Économisez 46,00 (10%)
-
Ajouter au panier
Quantité:
50 g
Grand volume ou format personnalisé
Prix (EUR)/ Each
414,00
Offre exceptionnelle en ligne
460.00 
Économisez 46,00 (10%)
Ajouter au panier
Oxyde de zinc, nanopoudre, 99 % (pureté du métal), Thermo Scientific Chemicals
Référence044533.18
Prix (EUR)/ Each
414,00
Offre exceptionnelle en ligne
460.00 
Économisez 46,00 (10%)
-
Ajouter au panier
Identifiants chimiques
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
Voir plus
Total Metal Impurities1% max.
Zinc oxide is incorporated in powders, ointments, and pastes. As Accelerator activator, pigment and reinforcing agent in rubber, ointments, pigment and mould-growth inhibitor in paints, uv absorber in plastics, ceramics, floor tile, glass, zinc salts, feed additive, dietary supplement, seed treatment cosmetics, photoconductor in office copying machines and in color photography, piezoelectric devices, artists' colorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only