5-phosphate de pyridoxal monohydraté, 99 %
5-phosphate de pyridoxal monohydraté, 99 %
5-phosphate de pyridoxal monohydraté, 99 %
5-phosphate de pyridoxal monohydraté, 99 %
Thermo Scientific Chemicals

5-phosphate de pyridoxal monohydraté, 99 %

CAS: 41468-25-1 | C8H10NO6P | 247.14 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 g
5 g
25 g
Conditionnement:
Flacon en verre
Référence 228170010
Prix (EUR)
33,49
Online Exclusive
40,40
Économisez 6,91 (17%)
Each
-
Ajouter au panier
Quantité:
1 g
Conditionnement:
Flacon en verre
Recurring order eligible. Learn more »
Grand volume ou format personnalisé
Prix (EUR)
33,49
Online Exclusive
40,40
Économisez 6,91 (17%)
Each
Ajouter au panier
Identifiants chimiques
CAS41468-25-1
IUPAC Name[(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Molecular FormulaC8H10NO6P
InChI KeyNGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILESCC1=NC=C(COP(O)(O)=O)C(C=O)=C1O
Voir plus
Appearance (Color)Off-white to yellow
Appearance (Form)Powder
HPLC>=98.5 %
Water=<10 %
Heavy metals (as Pb)=<20 ppm
Voir plus
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentation et téléchargements

Certificats

Safety Data Sheets

Partager la référence catalogue, le nom ou le lien.

1x1 image pixel for data collection