Acide tannique, réactif ACS, Thermo Scientific Chemicals
Acide tannique, réactif ACS, Thermo Scientific Chemicals
Acide tannique, réactif ACS, Thermo Scientific Chemicals
Acide tannique, réactif ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acide tannique, réactif ACS, Thermo Scientific Chemicals

CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
500 g
2,5kg
Conditionnement:
Flacon en plastique
Référence 419990025
Prix (EUR)
277,10
Online Exclusive
334,00
Économisez 56,90 (17%)
Each
Ajouter au panier
Quantité:
2,5kg
Conditionnement:
Flacon en plastique
Recurring order eligible. Learn more »
Grand volume ou format personnalisé
Prix (EUR)
277,10
Online Exclusive
334,00
Économisez 56,90 (17%)
Each
Ajouter au panier
Identifiants chimiques
CAS1401-55-4
IUPAC Name2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate
Molecular FormulaC76H52O46
InChI KeyLRBQNJMCXXYXIU-UHFFFAOYNA-N
SMILESOC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Voir plus
Sugars, dextrinPasses test
Zinc (Zn)=<50 ppm
Appearance (Color)Light brown
Appearance (Form)Powder
Heavy metals (as Pb)=<30 ppm
Voir plus
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentation et téléchargements

Certificats

Numéro de lotCertificate TypeDateCatalog Number(s)
A0470175Certificate of Analysis17 mai 2025419995000, 419990025
A0472588Certificate of Analysis17 mai 2025419995000, 419990025
A0466236Certificate of Analysis19 sept. 2024419995000, 419990025
A0465112Certificate of Analysis05 août 2024419995000, 419990025
A0459537Certificate of Analysis22 janv. 2024419995000, 419990025
5 résultats affichés, recherchez un certificat spécifique ci-dessus

Safety Data Sheets

Sécurité et manipulation

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H319 - Causes serious eye irritation

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear eye protection/ face protection

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P337 + P313 - If eye irritation persists: Get medical advice/attention

Partager la référence catalogue, le nom ou le lien.

1x1 image pixel for data collection