Limonine
Limonine
Limonine
Thermo Scientific Chemicals

Limonine

CAS: 1180-71-8 | C26H30O8 | 470.52 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
250 mg
1 g
Référence 465470010
Prix (EUR)
425,00
Online Exclusive
513,00
Économisez 88,00 (17%)
Each
-
Ajouter au panier
Quantité:
1 g
Grand volume ou format personnalisé
Prix (EUR)
425,00
Online Exclusive
513,00
Économisez 88,00 (17%)
Each
Ajouter au panier
Identifiants chimiques
CAS1180-71-8
IUPAC Name(1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
Molecular FormulaC26H30O8
InChI KeyKBDSLGBFQAGHBE-MSGMIQHVSA-N
SMILESCC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1
Voir plus
Infrared spectrumConforms
Loss on drying=<5 %
HPLC>=98.5 %
Appearance (Color)White to almost white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documentation et téléchargements

Certificats

Safety Data Sheets

Sécurité et manipulation

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Partager la référence catalogue, le nom ou le lien.