Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HHQNTGGFFV AVLVKKSSMP WNKRQPKLQG KSAETRESTQ LSPADLTEGK PTDPSKLESP SFTGTGDTEI AHATEDLENN GSKKDGVCGP PPSKKMKLFG FKEDPFVFIP EDDPLFPPIE KFYALDPSFP RMNLLTRTTE GKKRQLYMVS KELRNVLLNN SEKMKVINTG IKVWCRNNSG EEFDCAFRLA QEGIYTLYPF INSRIITVSM EDVKILLTQE NPFFRKLSSE TYSQAKDLAK GSIVLKYEPD SANPDALQCP IVLCGWRGKA SIRTFVPKNE RLHYLRMMGL EVLGEKKKEG VILTNESAAS TGQPDNDVTE GQRAGEPNSP DAEEANSPDV TAGCDPAGVH PPR
NSUN2 encodes a methyltransferase that catalyzes the intron-dependent formation of 5-methylcytosine at C34 of tRNA-leu(CAA). Maturation of cytoplasmic tRNAs includes splicing of introns, which are located 1 nucleotide 3-prime from the anticodon in all intron-containing tRNA genes. In tRNA-leu(CAA), the first position of the anticodon, C34, is converted to 5-methylcytosine, a modification necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA. Mutations of NSUN2 result in autosomal recessive 5 mental retardation (MRT5).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-methycytoisine methyltransferase; hTrm4; Misu; mRNA cytosine C(5)-methyltransferase; Myc-induced SUN domain-containing protein; Myc-induced SUN-domain-containing protein; NOL1/NOP2/Sun domain family member 2; NOL1/NOP2/Sun domain family, member 2; NOP2/Sun domain family, member 2; NOP2/Sun RNA methyltransferase family, member 2; RNA cytosine C(5)-methyltransferase NSUN2; Substrate of AIM1/Aurora kinase B; tRNA (cytosine-5-)-methyltransferase NSUN2; tRNA cytosine C(5)-methyltransferase; tRNA methyltransferase 4 homolog
Gene Aliases: MISU; MRT5; NSUN2; SAKI; TRM4
UniProt ID: (Human) Q08J23
Entrez Gene ID: (Human) 54888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support