Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
CHRAC1 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.[supplied by OMIM, Apr 2004].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHRAC-1; CHRAC-15; CHRC1; Chromatin accessibility complex 15 kDa protein; Chromatin accessibility complex protein 1; DNA polymerase epsilon subunit p15; histone-fold protein CHRAC15; HuCHRAC15
Gene Aliases: CHARC1; CHARC15; CHRAC-1; CHRAC-15; CHRAC1; CHRAC15; YCL1
UniProt ID: (Human) Q9NRG0
Entrez Gene ID: (Human) 54108
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support