Thermo Scientific Chemicals

Méthyl 4-amino-3-hydroxybenzoate, 98 %, Thermo Scientific Chemicals

Référence: L05687.04
2 g, Each
Thermo Scientific Chemicals

Méthyl 4-amino-3-hydroxybenzoate, 98 %, Thermo Scientific Chemicals

Référence: L05687.04
2 g, Each
Quantité
Référence: L05687.04
également connu sous le numéro L05687-04
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
63435-16-5
IUPAC Name
methyl 3-amino-4-hydroxybenzoate
Molecular Formula
C8H9NO3
InChI Key
VNQABZCSYCTZMS-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(O)C(N)=C1
Assay (GC)
≥97.5%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
117.0-123.0?C
Appearance (Color)
Pale yellow to yellow to brown
Form
Crystals or powder or crystalline powder

Description

Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Le méthyle 4-amino-3-hydroxybenzoate est utilisé comme matière première importante et intermédiaire utilisée dans la synthèse organique, les produits pharmaceutiques, les produits agrochimiques et les colorants.

Remarques
Incompatible avec les agents oxydants forts, les acides forts et les bases. Stockez-le dans un endroit frais. Garder le récipient bien fermé dans un endroit sec et bien ventilé.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention