Smart-Vue Pro Duo Modules 868Mhz
Product Image
Thermo Scientific™
Smart-Vue Pro Duo Modules 868Mhz
Une efficacité accrue grâce à la surveillance sans fil à capteurs multiples
Have Questions?
Référence SVPHWRMOD022
Prix (EUR)
-
Demander un devis
Ask our AI about this Product
Smart-Vue Pro Duo 868 Mhz Only. Delivered with two 3.6 V lithium batteries, one sensor mounting kit, one mounting holder, one antenna and one 1.5 m/5 ft. flat cable.
  • Bright 2.4 in. color touch screen with multiple views
  • Alarms locally with real -time audio-visual alarms on the modules
  • Audio visual alarms for local notification
  • Communication locally via Bluetooth and to the system via LoRaLow
  • AC power consumption through USB port access
  • Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
  • Choice of wired and wireless sensors with automatic sensor detection
  • User traceability with customized levels of user access with encrypted password protection
Spécifications
Autonomie de la batterie2 ans
Type de batterie3.6 V lithium battery
Certifications/ConformitéSmart-Vue Pro garantit la conformité avec la norme 21 CFR partie 11, les cGxP et les SOP.
ComprendTwo 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
Type d’affichageLCD
TypeModule
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Can other reducing agents other than DTT or BME be used to reduce proteins prior to electrophoresis? For example, what about TCEP (Tris Carboxy Ethyl Phosphene)?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documentation et téléchargements
Certificats
1 résultat affiché, recherchez un certificat spécifique ci-dessus
Partager la référence catalogue, le nom ou le lien.