Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
50131302 | Discontinued; replacement product is 50136167 |
50131250 | Discontinued; replacement product is 50136150 |
50131252 | Discontinued; replacement product is 50136151 |
50131296 | Discontinued; replacement product is 50136153 |
50131315 | Discontinued; replacement product is 50136169 |
50131317 | Discontinued; replacement product is 50136170 |
50131321 | Discontinued; replacement product is 50136171 |
50131323 | Discontinued; replacement product is 50136172 |
This item is no longer available and has been replaced by the next generation of Thermo Scientific™ Barnstead™ Water Purification Systems. The recommended replacement for this product is Barnstead™ GenPure™ xCAD Plus.
Gain flexibility and control with the compact remote dispenser in the Thermo Scientific™ Barnstead™ GenPure™ xCAD Water Purification System. This system delivers ultrapure water with consistent quality for the most demanding and sensitive applications. The remote dispenser can be wall- or bench-mounted. Ideal for molecular biology, microbiology and analytical chemistry applications, the system features integrated feed water monitoring, quick-connect cartridge replacement and exact dosing.
Integrated feed water monitoring:
UV Photo-oxidation 185/254 nm:
Ready to use:
Measurement of conductivity/resistivity:
Quick-connect cartridge replacement:
Exact dosing:
GLP-compliant documentation: