Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • p35 Antibodies

            Rockland

            p35 Polyclonal Antibody

            View all (10) p35 antibodies

            Promotions

            View available promotion(s)

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 600-401-ME9S
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            143.00
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite p35 Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            p35 Antibody in Immunohistochemistry (IHC)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            p35 Antibody (600-401-ME9S) in IHC

            Immunohistochemistry of Rabbit Anti-p35 Antibody. Tissue: Mouse Brain. Fixative: none. Antigen Retrieval: HIER. Primary Antibody: Anti-p35/Cdk5 Antibody at 1:100. Secondary Antibody: Ready-to-Use Anti-Rabbit. Counterstain: Hematoxylin. Results: p35 developed a strong nuclear and membranous staining of neurons and neutrophil in brain tissue.
            View Product

            p35 Antibody in Immunohistochemistry (IHC)
            p35 Antibody in Western Blot (WB)
            p35 Antibody in Western Blot (WB)

            Product Details

            600-401-ME9S

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000
            -

            Immunohistochemistry (IHC)

            1:100
            -

            ELISA (ELISA)

            1:10,000-1:50,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Anti-p35/Cdk5 antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a near N-terminal portion of mouse p35/Cdk5 conjugated to Keyhole Limpet Hemocyanin (KLH).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.96 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

            Contains

            0.01% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

            This affinity purified antibody is directed against mouse p35/Cdk5. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to rat,human, bovine, and Spermophilus citellus.

            Target Information

            The protein encoded by this gene (p35) is a neuron-specific activator of cyclin-dependent kinase 5 (CDK5); the activation of CDK5 is required for proper development of the central nervous system. The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form. The cleavage of p35 into p25 results in relocalization of the protein from the cell periphery to nuclear and perinuclear regions. P25 deregulates CDK5 activity by prolonging its activation and changing its cellular location. The p25 form accumulates in the brain neurons of patients with Alzheimer's disease. This accumulation correlates with an increase in CDK5 kinase activity, and may lead to aberrantly phosphorylated forms of the microtubule-associated protein tau, which contributes to Alzheimer's disease.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: CDK5 activator 1; Cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); neuronal CDK5 activator; regulatory partner for CDK5 kinase; tau protein kinase II 23kDa subunit; TPKII regulatory subunit

            View more View less

            Gene Aliases: CDK5P35; CDK5R; CDK5R1; D11Bwg0379e; NCK5A; p23; p25; p35; p35nck5a

            View more View less

            UniProt ID: (Human) Q15078, (Mouse) P61809

            View more View less

            Entrez Gene ID: (Human) 8851, (Mouse) 12569

            View more View less

            Function(s)

            protein kinase activity cyclin-dependent protein serine/threonine kinase activity calcium ion binding protein binding kinase activity cyclin-dependent protein kinase 5 activator activity protein kinase binding protein serine/threonine kinase activator activity cadherin binding ephrin receptor binding cytoskeletal protein binding kinase activator

            Process(es)

            regulation of cyclin-dependent protein serine/threonine kinase activity neuron migration neuron cell-cell adhesion G-protein coupled acetylcholine receptor signaling pathway axon guidance axonal fasciculation brain development cell proliferation embryo development regulation of macroautophagy peptidyl-serine phosphorylation peptidyl-threonine phosphorylation cerebellum development superior olivary nucleus maturation hippocampus development layer formation in cerebral cortex neuron differentiation negative regulation of axon extension neuron projection development regulation of actin cytoskeleton organization serine phosphorylation of STAT3 protein ionotropic glutamate receptor signaling pathway positive regulation of neuron apoptotic process regulation of neuron differentiation negative regulation of transcription, DNA-templated ephrin receptor signaling pathway rhythmic process regulation of dendritic spine morphogenesis regulation of microtubule cytoskeleton organization positive regulation of cell cycle arrest positive regulation of protein serine/threonine kinase activity positive regulation of protein targeting to membrane regulation of signal transduction by p53 class mediator protein phosphorylation cerebral cortex radially oriented cell migration positive regulation of protein kinase activity regulation of cell cycle
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0