Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Instrument Management

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CACNG5 Antibodies

            Alomone Labs, LTD

            CACNG5 (extracellular) Polyclonal Antibody

            View all (10) CACNG5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # ACC-115-25UL
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (HKD)
            5,015.00
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite CACNG5 (extracellular) Polyclonal Antibody

            • Antibody Testing Data (2)
            Application
            CACNG5 (extracellular) Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            CACNG5 (extracellular) Antibody (ACC-115-25UL) in ICC/IF

            Expression of CaV gamma 5 (CACNG5) in living rat PC12 cells - Cell surface detection of CaV gamma 5 (CACNG5) in intact living rat PC12 cells using Anti-CACNG5 (extracellular) Antibody (#ACC-115), (1:25) followed by goat Anti-rabbit-AlexaFluor-594 secondary Antibody . A. Extracellular staining (red) merged with live view of the cells. B. Extracellular staining merged with nuclear staining using DAPI as the c... View More
            View Product

            CACNG5 (extracellular) Antibody in Immunocytochemistry (ICC/IF)
            CACNG5 (extracellular) Antibody in Western Blot (WB)

            Product Details

            ACC-115-25UL

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:100
            -

            Immunocytochemistry (ICC/IF)

            1:25
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            (C)SESTVNVLKMIRS, corresponding to amino acid residues 87-99 of rat CACNG5, 1st extracellular loop
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            0.8 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.4, with 1% BSA

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

            Target Information

            L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: calcium channel, voltage-dependent, gamma subunit 5; Neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; Transmembrane AMPAR regulatory protein gamma-5; voltage dependent calcium channel gamma 5 subunit; Voltage-dependent calcium channel gamma-5 subunit

            View more View less

            Gene Aliases: CACNG5

            View more View less

            UniProt ID: (Human) Q9UF02, (Rat) Q8VHW8, (Mouse) Q8VHW4

            View more View less

            Entrez Gene ID: (Human) 27091, (Rat) 140726, (Mouse) 140723

            View more View less

            Function(s)

            voltage-gated calcium channel activity ion transmembrane transporter activity channel regulator activity voltage-gated ion channel activity calcium channel activity protein binding voltage-gated ion channel

            Process(es)

            transport transmission of nerve impulse cardiac conduction calcium ion transmembrane transport regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity ion transport calcium ion transport regulation of ion transmembrane transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            hong kong flag icon
            Hong Kong

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-cbcgw:80/100.66.73.215:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0