Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Betaine/GABA transporter 1; GABA transporter 1; gamma-aminobutyric acid (GABA-A) transporter 2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; Solute carrier family 6 member 12; vesicular GABA transporter
Gene Aliases: BGT1; Gabt2; Gat1; GAT2; RNU28927; Slc6a12; VGAT
UniProt ID: (Rat) P48056
Entrez Gene ID: (Mouse) 14411, (Rat) 50676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support