Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVFAFWKVFL ILSCLAGQVS VVQVTIPDGF VNVTVGSNVT LICIYTTTVA SREQLSIQWS FFHKKEMEPI SIYFSQGGQA VAIGQFKDRI TGSNDPGNAS ITISHMQPAD SGIYICDVNN PPDFLGQNQG ILNVSVLVKP SKPLCSVQGR PETGHTISLS CLSALGTPSP VYYWHKLEGR DIVPVKENFN PTTGILVIGN LTNFEQGYYQ CTAINRLGNS SCEIDLTSSH PEVGIIVGAL IGSLVGAAII ISVVCFARNK AKAKAKERNS KTIAELEPMT KINPRGESEA MPREDATQLE VTLPSSIHET GPDTIQEPDY EPKPTQEPAP EPAPGSEPMA VPDLDIELEL EPETQSELEP EPEPEPESEP GVVVEPLSED EKGVVKA
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell surface A33 antigen; Glycoprotein A34; MGC44287; V-set and immunoglobulin domain-containing protein 1
Gene Aliases: 1700062D20Rik; dJ889N15.1; GPA34; VSIG1
UniProt ID: (Human) Q86XK7
Entrez Gene ID: (Human) 340547
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support