Gold sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Quantity:
5 g
1 g
Catalog number 000827.06
also known as 000827-06
Price (JPY)/ Each
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSponge, -20 mesh
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only