Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis), Thermo Scientific Chemicals
Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold flake/sphere, APS 1-15 micron, 99.95% (metals basis), Thermo Scientific Chemicals

Quantity:
5 g
1 g
Catalog number 012453.06
also known as 012453-06
Price (JPY)/ Each
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGold flake/sphere
CAS7440-57-5
Recommended StorageAmbient temperatures
EINECS Number231-165-9
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only