Cerium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Cerium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Used as a pharmaceutical intermediate.
Quantity:
5 g
1 g
25 g
Catalog number 013641.06
also known as 013641-06
Price (JPY)/ Each
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7790-87-6
IUPAC Namecerium(3+) triiodide
Molecular FormulaCeI3
InChI KeyZEDZJUDTPVFRNB-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green to brown
FormPowder
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.1% max.
Cerium(III) iodide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only