Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals
Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.7g/10cm; Temper: soft; Formula: Pt:Ir; 80:20 wt%
Catalog number 039526.BS
Price (JPY)/ Each
-
Request A Quote
Quantity:
10 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum Iridium
ColorSilver to Gray
CAS7440-06-4
AppearanceSilver gray wire
Recommended StorageAmbient temperatures
View more
Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only