Thermo Scientific Chemicals

Malonaldehyde bis(dimethyl acetal), 99+%, Thermo Scientific Chemicals

Catalog number: 148610051
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Malonaldehyde bis(dimethyl acetal), 99+%, Thermo Scientific Chemicals

Catalog number: 148610051
5 kg, Plastic drum, Each
Quantity
Packaging

Chemical Identifiers

CAS
102-52-3
IUPAC Name
1,1,3,3-tetramethoxypropane
Molecular Formula
C7H16O4
InChI Key
XHTYQFMRBQUCPX-UHFFFAOYSA-N
SMILES
COC(CC(OC)OC)OC
Refractive index
1.4050 to 1.4090 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow-brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text