Thermo Scientific Chemicals

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 188450250
25 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 188450250
25 g, Each
Quantity

Chemical Identifiers

CAS
433-97-6
IUPAC Name
2-(trifluoromethyl)benzoic acid
Molecular Formula
C8H5F3O2
InChI Key
FBRJYBGLCHWYOE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1C(F)(F)F
Appearance (Color)
White to yellow-brown
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Form)
Powder
Melting point
106°C to 112°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text