2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, Thermo Scientific Chemicals
2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, Thermo Scientific Chemicals

Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 257690250
Price (JPY)/ Each
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
Molecular FormulaC6H2Br4O
InChI KeyNJQJGRGGIUNVAB-UHFFFAOYSA-N
SMILESBrC1=CC(Br)(Br)C=C(Br)C1=O
Molecular Weight (g/mol)409.70
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Appearance (Form)Powder
Titration Iodometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO ā€“ Research Use Only
1x1 image pixel for data collection