Thermo Scientific Chemicals

5-Chlorothiophene-2-boronic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 359010010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Chlorothiophene-2-boronic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 359010010
1 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
162607-18-3
IUPAC Name
(5-chlorothiophen-2-yl)boronic acid
Molecular Formula
C4H4BClO2S
InChI Key
JFCLNCVCDFUJPO-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(Cl)S1
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration with NaOH
97.0 to 103.0 %
Appearance (Color)
Off-white to light green to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text