Thermo Scientific Chemicals

Tetrabromophenolphthalein ethyl ester, pure

Catalog number: 420070250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabromophenolphthalein ethyl ester, pure

Catalog number: 420070250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
1176-74-5
IUPAC Name
potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular Formula
C22H13Br4KO4
InChI Key
WCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES
[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
Infrared spectrum
Conforms
Appearance (Form)
Powder
Appearance (Color)
Yellow to red
UV spectrophotometry
>=80.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text