Thermo Scientific Chemicals

Azasetron hydrochloride

Catalog number: 465072500
250 mg, Each
Thermo Scientific Chemicals

Azasetron hydrochloride

Catalog number: 465072500
250 mg, Each
Quantity

Chemical Identifiers

CAS
123040-16-4
IUPAC Name
hydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Molecular Formula
C17H21Cl2N3O3
InChI Key
DBMKBKPJYAHLQP-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
Appearance (Color)
White to beige
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
301.0°C to 303.0°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text