Thermo Scientific Chemicals

2,4,6-Triisopropylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A11280.22
100 g, Each
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A11280.22
100 g, Each
Quantity
Catalog number: A11280.22
also known as A11280-22
Price (JPY)

Chemical Identifiers

CAS
57199-00-5
IUPAC Name
2,4,6-tris(propan-2-yl)benzoyl chloride
Molecular Formula
C16H23ClO
InChI Key
OSKNTKJPGKHDHV-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(C(C)C)=C(C(Cl)=O)C(=C1)C(C)C
Form
Crystals or crystalline powder or fused solid
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Assay (GC)
≥98.0%
Melting Point (clear melt)
77-83?C
Identification (FTIR)
Conforms

Description

2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text