Thermo Scientific Chemicals

Isopropyl cyanoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A11887.14
25 g, Each
Thermo Scientific Chemicals

Isopropyl cyanoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A11887.14
25 g, Each
Quantity
Catalog number: A11887.14
also known as A11887-14
Price (JPY)

Chemical Identifiers

CAS
13361-30-3
IUPAC Name
propan-2-yl 2-cyanoacetate
Molecular Formula
C6H9NO2
InChI Key
BESQLCCRQYTQQI-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)CC#N
Refractive Index
1.4140-1.4190 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale pink
Assay (GC)
≥96.0%

Description

Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing KnÖevenagel reactions with aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text