Thermo Scientific Chemicals

3-Methylbenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A19568.18
50 g, Each
Thermo Scientific Chemicals

3-Methylbenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: A19568.18
50 g, Each
Quantity
Catalog number: A19568.18
also known as A19568-18
Price (JPY)

Chemical Identifiers

CAS
643-65-2
Form
Liquid
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5970-1.6000 @ 20?C

Description

3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text