You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search All
Aspire Free Products
Popular Product Areas
TaqMan Assays
Primary Antibodies
Secondary Antibodies
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
Methanol, Optima™ for HPLC, Fisher Chemical™
Element Search
Structure Search
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Fisher Chemical™
Methanol, Optima™ for HPLC, Fisher Chemical™
High purity mobile phase for HPLC
Technical Support
Customer Service
Catalog Number
Quantity
Packaging
A4544
4 L
Amber Soda Lime Glass
Try the chem dex
TM
web app
Catalog number
A4544
Price (JPY)
-
Contact Us ›
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Specifications
Absorbance
0.01 AU max. at 254nm, 0.11 AU max. at 230nm, 0.23 AU max. at 220nm, 1.00 AU max. at 205nm
Assay Percent Range
99.9% min by GC
CAS
67-56-1
ChEBI
CHEBI:17790
ChemAlert Storage Symbol
Red
View more
High purity mobile phase for HPLC
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Tested for HPLC gradient suitability
Mobile phase for analytical and preparative separations
Also meets ACS specifications
Intended for research use only
Please enter sku to load default questions
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link