Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™
Fisher Chemical™

Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™

High purity mobile phase for HPLC and extraction solvent
Catalog NumberQuantityPackaging
A46444 LAmber Soda Lime Glass
Catalog number A4644
Price (JPY)
-
Contact Us ›
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Specifications
Absorbance0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
Assay Percent RangeAssay: 99.9% min by GC
CAS67-63-0
ChEBICHEBI:17824
ChemAlert Storage SymbolRed
View more
Ask our AI about this Product
High purity mobile phase for HPLC and extraction solvent
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Also meets ACS specifications
  • Intended for research use only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link