Thermo Scientific Chemicals

4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25427.14
25 g, Each
Thermo Scientific Chemicals

4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25427.14
25 g, Each
Quantity
Catalog number: B25427.14
also known as B25427-14
Price (JPY)

Chemical Identifiers

CAS
3446-89-7
IUPAC Name
4-(methylsulfanyl)benzaldehyde
Molecular Formula
C8H8OS
InChI Key
QRVYABWJVXXOTN-UHFFFAOYSA-N
SMILES
CSC1=CC=C(C=O)C=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.6465-1.6515 @ 20?C
Form
Liquid
Free acid (titration)
≤1.5%

Description

4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text