1x1 image pixel for data collection
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™
Fisher BioReagents™

PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4, Fisher BioReagents™

Catalog NumberQuantity
BP2438-44L
Catalog number BP2438-4
Price (JPY)
-
Contact Us ›
Quantity:
4L
PBS, Phosphate Buffered Saline, 1X Solution, pH 7.4
A standard phosphate buffer used in many biomolecular procedures.
Specifications
Melting Point0°C
Packaging TypePoly Cube
Quantity4L
FormLiquid
pH7.3 to 7.5
Unit SizeEach
Contents & Storage
Room temperature

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection