Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Quantity:
250 g
500 g
Catalog number C-4320-50
also known as C/4320/50
Price (JPY)
-
Contact Us ›
Quantity:
250 g
Request bulk or custom format
Specifications
CAS7080-50-4
ColorWhite
Formula Weight281.69g/mol
IUPAC Namesodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
InChI KeyRNROOTWAJBXJBD-UHFFFAOYSA-M
View more
Ask our AI about this Product
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link