4-(Hydroxymethyl)benzeneboronic acid pinacol ester, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Hydroxymethyl)benzeneboronic acid pinacol ester, Thermo Scientific Chemicals
Quantity:
5 g
1 g
25 g
Catalog number H51123.06
also known as H51123-06
Price (JPY)/ Each
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Hydroxymethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Kimberly D.Grimes; Amol Gupte; Courtney C.Aldrich. Copper(II)-catalyzed conversion of aryl/heteroaryl boronic acids, boronates, and trifluoroborates into the corresponding azides: Substrate scope and limitations. Synthesis (Stuttg). 2010, (9),1441-1448