It is applied for desulfurization, decarburization extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied for desulfurization, decarburization extraction.
Solubility
Soluble in water.
Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
- Takashi Abe.; Irina Soloshonok.; Hajime Baba.; Akira Sekiya. Electrochemical fluorination of di- and tri-(2-hydroxyethyl)-substituted alkylamines. J. Fluorine Chem. 1999, 99 (1),51-57.
- D. Y. Chao.; W. J. Kuo.; N. H. Wang.; C. L. Lee.; K. H. Yang. Study of the synthesis and physical properties of fire-resistant polyurethane ionomers.J. Appl. Polym.Sc. 1998, 67 (1),19-26.