2,5-Dibromohydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Dibromohydroquinone, 97%, Thermo Scientific Chemicals
Quantity:
5 g
25 g
Catalog number H55453.06
also known as H55453-06
Price (JPY)/ Each
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14753-51-6
IUPAC Name2,5-dibromobenzene-1,4-diol
Molecular FormulaC6H4Br2O2
InChI KeyVALXCIRMSIFPFN-UHFFFAOYSA-N
SMILESOC1=CC(Br)=C(O)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
It is an important pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate.

Solubility
Soluble in methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
RUO – Research Use Only
Hayato Tsuji.; Chikahiko Mitsui.; Laurean Ilies.; Yoshiharu Sato.; Eiichi Nakamura. Synthesis and Properties of 2,3,6,7-Tetraarylbenzo[1,2-b:4,5-b‘]difurans as Hole-Transporting Material. J. Am. Chem. Soc. 2007, 129 (39), 11902-11903.