Malonyl dichloride, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonyl dichloride, 94%, Thermo Scientific Chemicals
Quantity:
100 g
25 g
Catalog number H59868.22
also known as H59868-22
Price (JPY)/ Each
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1663-67-8
IUPAC Namepropanedioyl dichloride
Molecular FormulaC3H2Cl2O2
InChI KeySXYFKXOFMCIXQW-UHFFFAOYSA-N
SMILESClC(=O)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay from Supplier's CofA≥92.0 to ≤108.0%
FormLiquid
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Decomposes in water.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Sarah J. Spessard.; Brian M. Stoltz. Progress toward the Synthesis of Garsubellin A and Related Phloroglucins: The Direct Diastereoselective Synthesis of the Bicyclo[3.3.1]nonane Core. Org. Lett. 2002, 4 (11),1943-1946.
  2. H.-G. Mack.; H. Oberhammer.; C.O.Della Védova. Gas-phase structure and conformational properties of malonyl dichloride, ClC(O)CH2C(O)Cl. J. Mol. Struct. 1995, 346, 51-61.