2-Hydroxy-4-nitropyridine, 96%, Thermo Scientific Chemicals
2-Hydroxy-4-nitropyridine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxy-4-nitropyridine, 96%, Thermo Scientific Chemicals

Quantity:
250 mg
1 g
Catalog number H66852.MD
also known as H66852-MD
Price (JPY)/ Each
-
Request A Quote
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS4487-51-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormSolid
Assay (HPLC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only