Thermo Scientific Chemicals

4-Aminobenzoic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J63428.22
100 g, Each
Thermo Scientific Chemicals

4-Aminobenzoic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: J63428.22
100 g, Each
Quantity
Catalog number: J63428.22
also known as J63428-22
Price (JPY)

Chemical Identifiers

CAS
555-06-6
IUPAC Name
sodium 4-aminobenzoate
Molecular Formula
C7H6NNaO2
InChI Key
XETSAYZRDCRPJY-UHFFFAOYSA-M
SMILES
[Na+].NC1=CC=C(C=C1)C([O-])=O
Form
Powder
Appearance (Color)
Pale yellow
Purity
≥97.5% (HPLC)

Description

4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzoic acid sodium salt is used as a Pharmaceutical intermediate.

Solubility
Soluble in water (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text