Gold nanoparticles, 20nm, carboxy-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold nanoparticles, 20nm, carboxy-functionalized, 3kDa PEGylated, OD50, 524nm absorption, Thermo Scientific Chemicals
Quantity:
1 mL
0.5 mL
Catalog number J67386.XF
also known as J67386-XF
Price (JPY)/ Each
-
Request A Quote
Quantity:
1 mL