Thermo Scientific Chemicals

1-Pentadecyne, 98%, Thermo Scientific Chemicals

Catalog number: L02540.04
2 g, Each
Thermo Scientific Chemicals

1-Pentadecyne, 98%, Thermo Scientific Chemicals

Catalog number: L02540.04
2 g, Each
Quantity
Catalog number: L02540.04
also known as L02540-04
Price (JPY)

Chemical Identifiers

CAS
765-13-9
GC/MS Analysis
Conforms
Refractive Index
1.442
Appearance (Color)
Colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

1-Pentadecyne is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecyne is used in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store under inert gas. Air sensitive. Avoid excessive heat and light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text