Thermo Scientific Chemicals

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19121.14
25 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%, Thermo Scientific Chemicals

Catalog number: L19121.14
25 g, Each
Quantity
Catalog number: L19121.14
also known as L19121-14
Price (JPY)

Chemical Identifiers

CAS
66399-30-2
IUPAC Name
1-(4-fluorophenyl)ethan-1-amine
Molecular Formula
C8H10FN
InChI Key
QGCLEUGNYRXBMZ-UHFFFAOYNA-N
SMILES
CC(N)C1=CC=C(F)C=C1
Form
Liquid
Optical Rotation
-33.0? ± 1.0? (neat)
Enantiomeric excess
≥98.5%
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text