The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography, disinfectants, wood preservatives
Solubility
Soluble in diethyl ether. Insoluble in water.
Notes
Store at room temperature. Reactive with oxidizing agents and strong bases.
RUO – Research Use Only
Jay A. Young. Mercury(II) Chloride. J. Chem. Educ. 2004, 81 (9),1260.
Kevin R Henke.; David Robertson.; Matthew K Krepps.; David A Atwood. Chemistry and stability of precipitates from aqueous solutions of 2,4,6-trimercaptotriazine, trisodium salt, nonahydrate (TMT-55) and mercury (II) chloride. Water Res.. 2000, 34 (11), 3005-3013.
Aromatic mercuration can be brought about by reaction with aryllithium compounds, e.g. from N,N-Diethylbenzamide, L08427: J. Org. Chem., 2728 (1988).
For a review of organomercurials in synthesis, see: Tetrahedron, 38, 1713 (1982).
Reagent for the cleavage of various sulfur compounds, such as dithioacetals; see 1,3-Dithiane, A10505.