Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Fisher Scientific™

Easy-Squeeze Wash Bottles, 6/Cs.

Easy to fill and ready to use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorChemical Label
028973BlueDeionized Water
0289712
also known as FIS-0289712
AssortedAssorted Solvents
0289713AssortedAssorted Cleaning
028979BlueSoap
0289711
also known as FIS-0289711
BlueWater
028974BlueDistilled Water
028977
also known as FIS-028977
GreenMethanol
028978NaturalSaline
0289710NaturalUniversal
028972NaturalSodium Hypochlorite (Bleach)
028975NaturalEthanol
028971
also known as FIS-028971
RedAcetone
028976YellowIsopropanol
Catalog number 028973
Price (KRW)
-
Color:
Blue
Chemical Label:
Deionized Water
Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Specifications
Capacity17 oz., 500mL
Closure ColorBlue
Closure MaterialPolypropylene
Closure Size38-415 mm
Closure TypePolypropylene Closure
Diameter (English)2.99 in.
Diameter (Metric)76 mm
Height (English)6.81 in.
Height (Metric)173 mm
IncludesPTFE membrane
Through the Closure DispensingNo
AutoclavableNot Autoclavable
Capacity (English)17 oz.
Capacity (Metric)500 mL
Certifications/ComplianceISO 13485:2003
Chemical LabelDeionized Water
Closure IncludedYes
ColorBlue
GraduatedNo
MaterialLDPE
MouthWide
Product TypeWash Bottle
ShapeCylindrical
Unit SizeCase of 6

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link