Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Fisher Scientific™

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConductivityResistivityConcentration or Composition Notes
0666424
also known as FISHERTOPSELLING-0666424-EACH
5 μS0.2 mOhmNIST Batch Tested
15077947
also known as FISHERTOPSELLING-15077947-EACH
5 μS0.5 mOhmNIST Individual Bottle Tested
15077945
also known as FISHERTOPSELLING-15077945-EACH
1 μS1.0 mOhmNIST Batch Tested
15077946
also known as FISHERTOPSELLING-15077946-EACH
1 μS1.0 mOhmNIST Individual Bottle Tested
Catalog number 0666424
also known as FISHERTOPSELLING-0666424-EACH
Price (KRW)
201,000
Each
Add to cart
Conductivity:
5 μS
Resistivity:
0.2 mOhm
Concentration or Composition Notes:
NIST Batch Tested
Price (KRW)
201,000
Each
Add to cart
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Specifications
Chemical Name or MaterialConductivity Standard
ColorClear
Concentration or Composition NotesNIST Batch Tested
Conductivity5 μS
DescriptionNIST Batch Tested, Resistivity: 0.2 mΩ
For Use With (Equipment)Conductivity Meter
PackagingBottle
Quantity1 x 473 mL (16 oz.)
Resistivity0.2 mOhm
SolubilitySoluble in water
Solution TypeCalibration Solution
Traceability to NISTNIST Batch Tested
TypeConductivity Standard
Certifications/ComplianceISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
TechniqueCalibration of Conductivity Instrument
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link