Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
07250106 also known as FISHERMIS-07250106-1000/PK | 1 L |
07250102 also known as FISHERMIS-07250102-EACH | 100 mL |
07250107 also known as FISHERMIS-07250107-EACH | 2000 mL |
These volumetric flasks provide precise volume measurement. The necks are tooled for standard taper glass stoppers. The graduation line is sharp and permanent and large blue block letters make the inscription easy to read. These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks.
Share catalog number, name or link