Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Tolerance |
---|---|---|
082612D also known as FISHERMIS-082612D-EACH | 90 mm | ±0.03 mL |
082612A also known as FISHERMIS-082612A-250/PK | 100 mm | ±0.02 mL |
Made from Code 7740 borosilicate glass. The 100 and 90 mm sizes are available with both ends open or with one end open and one closed. All tubes have a 0.2 mm to 0.25 mm wall range.
Share catalog number, name or link