Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) |
---|---|
086703C also known as FIS-086703C | 76 mm |
086703BB also known as FISHERMIS-086703BB-25/PK | 16 mm |
086703A also known as FISHERMIS-086703A-EACH | 45 mm |
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
Do not tie knots in tubing; use closures
Share catalog number, name or link