1x1 image pixel for data collection
MaximaDry™ Diaphragm Vacuum Pump
MaximaDry™ Diaphragm Vacuum Pump
Fisher Scientific™

MaximaDry™ Diaphragm Vacuum Pump

Clean, reliable, oil-free vacuum pumps are ideal for low to high boiling point rotary evaporation and distillation, filtration, blotting, and solid phase extraction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelFlow Rate
1388022MaximaDry™ 35 to 1.5; 115V35 L/min.
1388018MaximaDry™ 20 to 6 1;15V20 L/min.
1388014MaximaDry™ 20 to 75; 115V20 L/min.
1388020MaximaDry™ 35 to 6; 115V35 L/min.
1388016MaximaDry™ 40 to 65; 115V40 L/min.
Catalog number 1388022
Price (KRW)
-
Model:
MaximaDry™ 35 to 1.5; 115V
Flow Rate:
35 L/min.

Clean, reliable, oil-free vacuum pumps are ideal for high boiling point rotary evaporation and distillion, vacuum concentration, and larger vacuum ovens (>0.5 cu.ft.)

Quiet, reliable operation without the hazards of handling pump oil or contaminated water. Excellent replacement for rotary vane pumps and water aspirators, especially in applications where chemical vapors are encountered.

  • Withstands aggressive gases and vapors — can be used with aggressive solvents without the risk of damaging the pump
  • Single-piece PTFE coated diaphragm and molded PTFE pump heads provide chemical resistance
  • High-flow, multiport valves are made from tough FFPM fluoropolymer to withstand pumping heavy solvent vapors and liquid condensate without a loss in performance
  • Fan-cooled (TEFC) motor is rated for continuous operation and never needs lubrication
  • Compact design fits easily on a benchtop or in a fume hood
  • Setup and disassembly are quick and easy and do not require any special tools
  • Available in either 115 V/60 Hz or 230 V/50 Hz

Specifications
Max. Vacuum1.5 torr
Electrical Requirements115 V 60 Hz
Dimensions (L x W x H)16 x 7 x 9 in. (40.6 x 17.8 x 23 cm)
ModelMaximaDry™ 35 to 1.5; 115V
Flow Rate35 L/min.
Frequency60 Hz
Pump TypeDiaphragm Vacuum Pump
Enclosure RatingIP44
ApplicationsVacuum Concentration
Maximum Pressure15 psig
Voltage115 V
Unit SizeEach

Share catalog number, name or link