Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Fisher Scientific™

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™

Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConductivityResistivityConcentration or Composition Notes
15077946
also known as FISHERTOPSELLING-15077946-EACH
1 μS1.0 mOhmNIST Individual Bottle Tested
0666424
also known as FISHERTOPSELLING-0666424-EACH
5 μS0.2 mOhmNIST Batch Tested
15077947
also known as FISHERTOPSELLING-15077947-EACH
5 μS0.5 mOhmNIST Individual Bottle Tested
15077945
also known as FISHERTOPSELLING-15077945-EACH
1 μS1.0 mOhmNIST Batch Tested
Catalog number 15077946
also known as FISHERTOPSELLING-15077946-EACH
Price (KRW)
321,000
Each
Add to cart
Conductivity:
1 μS
Resistivity:
1.0 mOhm
Concentration or Composition Notes:
NIST Individual Bottle Tested
Delivery information
Standard Products - 1-2 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 2-3 business days for 2nd-tier cities, and 4 business days for 3rd-tier cities and remote areas.
Air Freight Restricted Products - 2-4 business days for Beijing, Shanghai, Guangzhou, Shenzhen and provincial capitals, 3-5 business days for 2nd-tier cities, and 6-10 business days for 3rd-tier cities and remote areas.
Supply Center: Delivered from the Supply Center closest to you.
Price (KRW)
321,000
Each
Add to cart
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Specifications
Chemical Name or MaterialConductivity Standard
ColorClear
Concentration or Composition NotesNIST Individual Bottle Tested
Conductivity1 μS
DescriptionNIST Individual Bottle Tested, Resistivity: 1.0 mΩ
For Use With (Equipment)Conductivity Meter
PackagingBottle
Quantity1 x 473 mL (16 oz.)
Resistivity1.0 mOhm
SolubilitySoluble in water
Solution TypeCalibration Solution
Traceability to NISTNIST Individual Bottle Tested
TypeConductivity Standard
Certifications/ComplianceISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
TechniqueCalibration of Conductivity Instrument
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link