1x1 image pixel for data collection
Mitcool 1700 MP Cleanroom Lab Coats
Mitcool 1700 MP Cleanroom Lab Coats
Mitcool 1700 MP Cleanroom Lab Coats
Mitcool 1700 MP Cleanroom Lab Coats
Mitcool 1700 MP Cleanroom Lab Coats
Mitcool 1700 MP Cleanroom Lab Coats
MP lab coats are constructed of a non-woven, microporous, coated material and provide protection in controlled environments.
Have Questions?
Catalog NumberdimSize
19546403
also known as FIS-19546403
Catalog number 19546403
also known as FIS-19546403
Price (KRW)
-
Ask our AI about this Product
MP lab coats are constructed of a non-woven, microporous, coated material and provide protection in controlled environments.
  • Bonded polypropylene with laminate
  • Liquid chemical resistance
  • Comfortable, yet durable
  • Low particle shedding
  • Disposable
Specifications
Product TypeControlled Environments Lab Coat
ColorWhite
Closure TypeZipper
Cuff StyleElastic
DisposableSingle-use
Seam TypeBound
Certifications/ComplianceASTM F903-96, ASTM F1407-96
ConstructionPolypropylene, Laminate Binding Material
Unit SizeCase of 25
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection