Humidity/Slide Moisture Chamber
Humidity/Slide Moisture Chamber
Fisher Scientific™
Humidity/Slide Moisture Chamber
Ideal for both immunostaining and diagnostic reactions performed on slides
Have Questions?
Catalog number 23769522
also known as FISHERTOPSELLING-23769522-EACH
Price (KRW)
-
Ideal for both immunostaining and diagnostic reactions performed on slides
  • All units divided into 10 compartments that separated by lid barrier
  • Chambers are made of heavy-duty polystyrene
  • Air tight sealing keep moisture inside
  • Stackable boxes enables easy storage
  • Black in color
Specifications
TypeSlide Moisture Chamber
ColorBlack
MaterialHeavy Duty Polystyrene
For Use With (Application)Both immunostaining and diagnostic reactions performed on slides
IncludesLid barrier between each slide
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link